Assay Details
| Gene Symbol: toe-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026872.4 | NP_001022043.1 | 2 - 3 | 373 | 65 | |
| NM_001383867.1 | 1 - 2 | 962 | 65 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

toe-2| Gene Symbol: toe-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026872.4 | NP_001022043.1 | 2 - 3 | 373 | 65 | |
| NM_001383867.1 | 1 - 2 | 962 | 65 | |||
Set Membership: |
|