Assay Details
| Gene Symbol: tra-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026783.6 | NP_001021954.1 | 3 - 4 | 388 | 80 | |
| NM_063025.9 | NP_495426.1 | 3 - 4 | 388 | 80 | ||
| GenBank mRNA | M91371.1 | - | 3 - 4 | 424 | 80 | |
| S42187.1 | - | 3 - 4 | 424 | 80 | ||
More Information
Set Membership: |
|

tra-2
Genomic Map