Assay Details
| Gene Symbol: pgam-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063192.5 | NP_495593.2 | 1 - 2 | 199 | 123 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pgam-5| Gene Symbol: pgam-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063192.5 | NP_495593.2 | 1 - 2 | 199 | 123 | |
Set Membership: |
|