Assay Details
| Gene Symbol: jun-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027195.4 | NP_001022366.1 | 2 - 3 | 321 | 94 | |
| NM_001027196.5 | NP_001022367.1 | 2 - 3 | 186 | 94 | ||
| GenBank mRNA | EU553920.1 | - | 2 - 3 | 259 | 94 | |
| Gene Symbol: C16D2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381554.1 | NP_001367159.1 | 2 - 3 | 344 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381554.1 transcripts.
Set Membership: |
|

jun-1
Genomic Map