Assay Details
| Gene Symbol: fzr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063674.6 | NP_496075.1 | 6 - 7 | 2120 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fzr-1| Gene Symbol: fzr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063674.6 | NP_496075.1 | 6 - 7 | 2120 | 68 | |
Set Membership: |
|