Assay Details
| Gene Symbol: F54D5.12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064064.9 | NP_496465.1 | 1 - 2 | 199 | 101 | |
| Gene Symbol: gtf-2E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064065.9 | NP_496466.1 | 5 - 6 | 1269 | 101 | |
More Information
Set Membership: |
|

F54D5.12
Genomic Map