Assay Details
| Gene Symbol: F54D5.12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064064.9 | NP_496465.1 | 3 - 4 | 1282 | 105 | |
| Gene Symbol: gtf-2E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064065.9 | NP_496466.1 | 7 - 8 | 2352 | 105 | |
More Information
Set Membership: |
|

F54D5.12
Genomic Map