Assay Details
| Gene Symbol: xrn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064544.6 | NP_496945.3 | 1 - 2 | 294 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

xrn-1| Gene Symbol: xrn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064544.6 | NP_496945.3 | 1 - 2 | 294 | 87 | |
Set Membership: |
|