Assay Details
| Gene Symbol: Y53G8AM.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065281.7 | NP_497682.2 | 7 - 8 | 2536 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Y53G8AM.8| Gene Symbol: Y53G8AM.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065281.7 | NP_497682.2 | 7 - 8 | 2536 | 64 | |
Set Membership: |
|