Assay Details
| Gene Symbol: dcn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001384069.1 | 2 - 3 | 336 | 126 | ||
| NM_065465.6 | NP_497866.2 | 3 - 4 | 493 | 126 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dcn-1| Gene Symbol: dcn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001384069.1 | 2 - 3 | 336 | 126 | ||
| NM_065465.6 | NP_497866.2 | 3 - 4 | 493 | 126 | ||
Set Membership: |
|