Assay Details
| Gene Symbol: hse-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065475.7 | NP_497876.2 | 2 - 3 | 463 | 76 | |
| GenBank mRNA | AY552607.1 | - | 2 - 3 | 455 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hse-5| Gene Symbol: hse-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065475.7 | NP_497876.2 | 2 - 3 | 463 | 76 | |
| GenBank mRNA | AY552607.1 | - | 2 - 3 | 455 | 76 | |
Set Membership: |
|