Assay Details
| Gene Symbol: hse-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065475.7 | NP_497876.2 | 3 - 4 | 560 | 123 | |
| GenBank mRNA | AY552607.1 | - | 3 - 4 | 552 | 123 | |
More Information
Set Membership: |
|

hse-5
Genomic Map