Assay Details
| Gene Symbol: cdh-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330837.3 | NP_001317871.1 | 5 - 6 | 1866 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cdh-4| Gene Symbol: cdh-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330837.3 | NP_001317871.1 | 5 - 6 | 1866 | 128 | |
Set Membership: |
|