Assay Details
| Gene Symbol: tmem-131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065658.8 | NP_498059.2 | 3 - 4 | 539 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tmem-131| Gene Symbol: tmem-131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065658.8 | NP_498059.2 | 3 - 4 | 539 | 124 | |
Set Membership: |
|