Assay Details
| Gene Symbol: bas-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025979.4 | NP_001021150.1 | 1 - 2 | 141 | 109 | |
| NM_001025980.5 | NP_001021151.1 | 1 - 2 | 141 | 109 | ||
More Information
Set Membership: |
|

bas-1
Genomic Map