Assay Details
| Gene Symbol: bas-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025979.4 | NP_001021150.1 | 3 - 4 | 814 | 75 | |
| NM_001025980.5 | NP_001021151.1 | 4 - 5 | 841 | 75 | ||
More Information
Set Membership: |
|

bas-1
Genomic Map