Assay Details
| Gene Symbol: tdo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065883.5 | NP_498284.1 | 5 - 6 | 842 | 75 | |
| NR_136366.1 | 4 - 5 | 1064 | 75 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tdo-2| Gene Symbol: tdo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065883.5 | NP_498284.1 | 5 - 6 | 842 | 75 | |
| NR_136366.1 | 4 - 5 | 1064 | 75 | |||
Set Membership: |
|