Assay Details
| Gene Symbol: tdo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047382.6 | NP_001040847.1 | 1 - 2 | 102 | 108 | |
| NM_065883.5 | NP_498284.1 | 6 - 7 | 979 | 108 | ||
| NR_136366.1 | 5 - 6 | 1201 | 108 | |||
More Information
Set Membership: |
|

tdo-2
Genomic Map