Assay Details
| Gene Symbol: ger-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066139.5 | NP_498540.1 | 1 - 2 | 86 | 73 | |
| GenBank mRNA | AM231686.1 | - | 1 - 2 | 49 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ger-1| Gene Symbol: ger-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066139.5 | NP_498540.1 | 1 - 2 | 86 | 73 | |
| GenBank mRNA | AM231686.1 | - | 1 - 2 | 49 | 73 | |
Set Membership: |
|