Assay Details
| Gene Symbol: ger-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066139.5 | NP_498540.1 | 2 - 3 | 181 | 130 | |
| GenBank mRNA | AM231686.1 | - | 2 - 3 | 144 | 130 | |
More Information
Set Membership: |
|

ger-1
Genomic Map