Assay Details
| Gene Symbol: psd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025956.5 | NP_001021127.1 | 3 - 4 | 847 | 85 | |
| NM_001025957.5 | NP_001021128.1 | 4 - 5 | 838 | 85 | ||
More Information
Set Membership: |
|

psd-1
Genomic Map