Assay Details
| Gene Symbol: gos-28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066220.5 | NP_498621.1 | 1 - 2 | 119 | 94 | |
| Gene Symbol: F08F8.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066219.8 | NP_498620.1 | 1 - 2 | 119 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_066219.8 transcripts.
Set Membership: |
|

gos-28
Genomic Map