Assay Details
| Gene Symbol: lgc-50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066236.5 | NP_498637.3 | 7 - 8 | 1045 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lgc-50| Gene Symbol: lgc-50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066236.5 | NP_498637.3 | 7 - 8 | 1045 | 81 | |
Set Membership: |
|