Assay Details
| Gene Symbol: clk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382917.1 | 9 - 10 | 1831 | 101 | ||
| GenBank mRNA | AF400665.1 | - | 1 - 2 | 161 | 101 | |
| AJ320257.1 | - | 1 - 2 | 155 | 101 | ||
More Information
Set Membership: |
|

clk-2
Genomic Map