Assay Details
| Gene Symbol: clk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382917.1 | 12 - 13 | 2504 | 110 | ||
| GenBank mRNA | AF400665.1 | - | 4 - 5 | 834 | 110 | |
| AJ320257.1 | - | 4 - 5 | 828 | 110 | ||
More Information
Set Membership: |
|

clk-2
Genomic Map