Assay Details
| Gene Symbol: cux-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066250.8 | NP_498651.2 | 7 - 8 | 1457 | 141 | |
| Gene Symbol: clk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382917.1 | 7 - 8 | 1497 | 141 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001382917.1 transcripts.
Set Membership: |
|

cux-7
Genomic Map