Assay Details
| Gene Symbol: lig-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379799.2 | NP_001367462.1 | 10 - 11 | 2040 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lig-4| Gene Symbol: lig-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379799.2 | NP_001367462.1 | 10 - 11 | 2040 | 100 | |
Set Membership: |
|