Assay Details
| Gene Symbol: grp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066363.4 | NP_498764.1 | 4 - 5 | 695 | 148 | |
| Gene Symbol: idi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382931.1 | 4 - 5 | 708 | 148 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001382931.1 transcripts.
Set Membership: |
|

grp-1
Genomic Map