Assay Details
| Gene Symbol: mut-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066704.9 | NP_499105.1 | 8 - 9 | 2708 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mut-7| Gene Symbol: mut-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066704.9 | NP_499105.1 | 8 - 9 | 2708 | 88 | |
Set Membership: |
|