Assay Details
| Gene Symbol: gtf-2H2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066838.6 | NP_499239.1 | 8 - 9 | 821 | 90 | |
| Gene Symbol: hal-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379881.2 | NP_001367068.1 | 8 - 9 | 835 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001379881.2 transcripts.
Set Membership: |
|

gtf-2H2C
Genomic Map