Assay Details
| Gene Symbol: mua-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027503.4 | NP_001022674.1 | 23 - 24 | 4822 | 64 | |
| NM_001027504.3 | NP_001022675.1 | 13 - 14 | 3462 | 64 | ||
| GenBank mRNA | AF139060.1 | - | 23 - 24 | 4822 | 64 | |
More Information
Set Membership: |
|

mua-3
Genomic Map