Assay Details
| Gene Symbol: mua-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027503.4 | NP_001022674.1 | 25 - 26 | 5258 | 153 | |
| NM_001027504.3 | NP_001022675.1 | 15 - 16 | 3898 | 153 | ||
| GenBank mRNA | AF139060.1 | - | 25 - 26 | 5258 | 153 | |
More Information
Set Membership: |
|

mua-3
Genomic Map