Assay Details
| Gene Symbol: dpy-28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066978.5 | NP_499379.2 | 2 - 3 | 364 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpy-28| Gene Symbol: dpy-28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066978.5 | NP_499379.2 | 2 - 3 | 364 | 103 | |
Set Membership: |
|