Assay Details
| Gene Symbol: tin-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382999.1 | 1 - 2 | 56 | 60 | ||
| GenBank mRNA | AF150094.1 | - | 1 - 2 | 40 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tin-10| Gene Symbol: tin-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382999.1 | 1 - 2 | 56 | 60 | ||
| GenBank mRNA | AF150094.1 | - | 1 - 2 | 40 | 60 | |
Set Membership: |
|