Assay Details
| Gene Symbol: crh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027688.5 | NP_001022859.1 | 5 - 6 | 762 | 82 | |
| NM_001027689.3 | NP_001022860.1 | 5 - 6 | 666 | 82 | ||
| NM_001027690.3 | NP_001022861.1 | 5 - 6 | 656 | 82 | ||
| NM_001027691.4 | NP_001022862.1 | 5 - 6 | 778 | 82 | ||
| NM_001027692.4 | NP_001022863.1 | 5 - 6 | 663 | 82 | ||
| NM_001268206.3 | NP_001255135.1 | 4 - 5 | 969 | 82 | ||
| NM_001268207.2 | NP_001255136.1 | 2 - 3 | 518 | 82 | ||
| GenBank mRNA | AB081595.1 | - | 5 - 6 | 663 | 82 | |
| AB081596.1 | - | 5 - 6 | 745 | 82 | ||
More Information
Set Membership: |
|

crh-1
Genomic Map