Assay Details
| Gene Symbol: crh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027688.5 | NP_001022859.1 | 1 - 2 | 175 | 69 | |
| NM_001027689.3 | NP_001022860.1 | 1 - 2 | 73 | 69 | ||
| GenBank mRNA | AB081596.1 | - | 1 - 2 | 158 | 69 | |
More Information
Set Membership: |
|

crh-1
Genomic Map