Assay Details
| Gene Symbol: arx-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001368436.4 | NP_001355516.1 | 3 - 4 | 718 | 93 | |
| NM_067169.7 | NP_499570.1 | 3 - 4 | 718 | 93 | ||
More Information
Set Membership: |
|

arx-3
Genomic Map