Assay Details
| Gene Symbol: osm-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067184.3 | NP_499585.1 | 9 - 10 | 1769 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

osm-12| Gene Symbol: osm-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067184.3 | NP_499585.1 | 9 - 10 | 1769 | 69 | |
Set Membership: |
|