Assay Details
| Gene Symbol: scav-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067224.7 | NP_499625.2 | 1 - 2 | 128 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

scav-3| Gene Symbol: scav-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067224.7 | NP_499625.2 | 1 - 2 | 128 | 97 | |
Set Membership: |
|