Assay Details
| Gene Symbol: aass-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307729.4 | NP_001294658.1 | 3 - 4 | 839 | 82 | |
| NM_067483.5 | NP_499884.1 | 3 - 4 | 868 | 82 | ||
More Information
Set Membership: |
|

aass-1
Genomic Map