Assay Details
| Gene Symbol: F18F11.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067505.5 | NP_499906.3 | 7 - 8 | 1530 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F18F11.4| Gene Symbol: F18F11.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067505.5 | NP_499906.3 | 7 - 8 | 1530 | 62 | |
Set Membership: |
|