Assay Details
| Gene Symbol: spl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067512.8 | NP_499913.1 | 2 - 3 | 191 | 128 | |
| GenBank mRNA | AF144639.1 | - | 2 - 3 | 185 | 128 | |
More Information
Set Membership: |
|

spl-1
Genomic Map