Assay Details
| Gene Symbol: pam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028038.7 | NP_001023209.1 | 6 - 7 | 1673 | 105 | |
| NM_001028039.3 | NP_001023210.1 | 5 - 6 | 1173 | 105 | ||
More Information
Set Membership: |
|

pam-1
Genomic Map