Assay Details
| Gene Symbol: dcaf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069324.6 | NP_501725.2 | 6 - 7 | 1022 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dcaf-1| Gene Symbol: dcaf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069324.6 | NP_501725.2 | 6 - 7 | 1022 | 129 | |
Set Membership: |
|