Assay Details
| Gene Symbol: dct-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047534.5 | NP_001040999.2 | 3 - 4 | 486 | 110 | |
| NM_001268500.2 | NP_001255429.1 | 3 - 4 | 503 | 110 | ||
More Information
Set Membership: |
|

dct-15
Genomic Map