Assay Details
| Gene Symbol: zim-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136397.4 | NP_001129869.1 | 2 - 3 | 831 | 86 | |
| NM_069547.4 | NP_501948.2 | 2 - 3 | 829 | 86 | ||
| Gene Symbol: zim-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383192.1 | 2 - 3 | 831 | 86 | ||
| Gene Symbol: zim-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307056.3 | NP_001293985.1 | 2 - 3 | 824 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001307056.3, NM_001383192.1 transcripts.
Set Membership: |
|

zim-1
Genomic Map