Assay Details
| Gene Symbol: zim-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136397.4 | NP_001129869.1 | 5 - 6 | 1588 | 75 | |
| NM_001276847.3 | NP_001263776.1 | 2 - 3 | 440 | 75 | ||
| NM_001276848.3 | NP_001263777.1 | 2 - 3 | 440 | 75 | ||
| NM_069547.4 | NP_501948.2 | 5 - 6 | 1586 | 75 | ||
More Information
Set Membership: |
|

zim-1
Genomic Map