Assay Details
| Gene Symbol: zim-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136397.4 | NP_001129869.1 | 9 - 10 | 3056 | 135 | |
| Gene Symbol: zim-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383192.1 | 5 - 6 | 2036 | 135 | ||
More Information
Set Membership: |
|

zim-1
Genomic Map