Assay Details
| Gene Symbol: glt-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067543.4 | NP_499944.3 | 8 - 9 | 1444 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

glt-7| Gene Symbol: glt-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067543.4 | NP_499944.3 | 8 - 9 | 1444 | 65 | |
Set Membership: |
|