Assay Details
| Gene Symbol: gex-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_067548.5 | NP_499949.2 | 10 - 11 | 3472 | 65 | |
| GenBank mRNA | AB073209.1 | - | 10 - 11 | 3494 | 65 | |
More Information
Set Membership: |
|

gex-2
Genomic Map